Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nuclear Antigen Rabbit anti-Human, Primate, Clone: 235-1R, Novus Biologicals™

Rabbit Monoclonal Antibody
$275.50 - $529.00
Specifications
Antigen | Nuclear Antigen |
---|---|
Clone | 235-1R |
Concentration | 0.2 mg/ml |
Dilution | Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1000-2000 ug/500 ug protein lysate, Immunohistochemistry-Frozen 1-2 ug/ml |
Applications | Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) |
Description
Nuclear Antigen Monoclonal antibody specifically detects Nuclear Antigen in Human, Primate samples. It is validated for Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen)Specifications
Nuclear Antigen | |
0.2 mg/ml | |
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
Unconjugated | |
Rabbit | |
Human, Primate | |
Nuclei, Nucleus | |
IgG κ | |
Protein A or G purified |
235-1R | |
Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1000-2000 ug/500 ug protein lysate, Immunohistochemistry-Frozen 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
Nuclei of human myeloid leukemia biopsy cells | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title