Learn More
Invitrogen™ Nuclear Matrix Protein p84 Polyclonal Antibody

Description
Immunogen sequence: KILMGNEELT RLWNLCPDNM EACKSETREH MPTLEEFFEE AIEQADPENM VENEYKAVNN SNYGWRALRL LARRSPHFFQ PTNQQFKSLP EYLENMVIKL AKELPPPSEE IKTGEDEDEE DNDALLK Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 88%.
Specifications
Specifications
| Antigen | Nuclear Matrix Protein p84 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.20 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Thoc1 |
| Gene Accession No. | P59924, Q8R3N6, Q96FV9 |
| Gene Alias | 3110002N20Rik; AW107452; HPR1; hTREX84; I79_014746; LRRGT00070; NMP-84; Nuclear matrix protein p84; P84; P84N5; THO complex 1; THO complex subunit 1; THO complex subunit 1-like protein; tho1; Thoc1 |
| Gene Symbols | Thoc1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.