Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nuclear Pore Complex Protein Nsp1p Antibody (32D6), Novus Biologicals™
SDP

Catalog No. NB018366 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018366 0.125 mL
NBP105227 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018366 Supplier Novus Biologicals Supplier No. NBP1052270.125ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nuclear Pore Complex Protein Nsp1p Monoclonal specifically detects Nuclear Pore Complex Protein Nsp1p in Yeast samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen Nuclear Pore Complex Protein Nsp1p
Applications Immunofluorescence, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 32D6
Conjugate Unconjugated
Dilution Western Blot 1:10000-1:50000, Immunohistochemistry 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:1000-1:10000
Formulation Tissue culture supernatant with 5mM Sodium Azide
Gene Alias 107 kDa nucleoporin, NPC proteins, Nuclear pore complex protein Nup107, Nucleoporin 107kDa, Nucleoporin Nup107, NUP 107, NUP 84, NUP107, NUP153, NUP155, NUP84, NUP98
Host Species Mouse
Immunogen Yeast nuclear preparations
Molecular Weight of Antigen 86.5 kDa
Purification Method Tissue culture supernatant
Quantity 0.125 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Yeast
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.