Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Magna Nuclear RIP™ (Native) Nuclear RNA-Binding Protein Immunoprecipitation Kit

Catalog No. 1710522 Shop All MilliporeSigma Products
Click to view available options
Quantity:
24 Assays

Includes

  • Nuclei Isolation Buffer
  • RIP Lysis Buffer
  • Protein A/G Magnetic Beads
  • RIP Lysis Buffer
  • 0.5M EDTA
  • DNase I (RNase Free) 2U/μL
  • DNase I Reaction Buffer
  • Protease Inhibitor Cocktail III, Animal Free
  • RNAse Inhibitor

Requires

  • Antibody of interest for RNA-binding protein immunoprecipitation (RIP)
  • Negative Control Antibody
  • Precipitation Carrier (e.g. Pellet Paint™ Co-Precipitant or linear Acrylamide (5mg/mL) if samples are intended for Next Gen Sequence library preparation
  • Reagents for qRT-PCR Analysis and/or Reagents for RNA-Seq Library Construction (optional)

Specially designed to allow the discovery and analysis of a variety of chromatin associated RNAs such as long non-coding RNAs, enhancer RNAs and miRNAs

  • Generates cross-linked chromatin to allow analysis of a variety of chromatin-associated RNAs
  • Flexible, scalable input requirements: Recover RNA from millions of cells or as few as 100,000 cells (native)
  • Magnetic protein A/G bead blend and optimized buffer system results in low backgrounds and high signal-to-noise ratios
  • Suitable for analysis by RT-qPCR or RIP-seq
  • Complete set of reagents and detailed protocol to enable first-time success
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store components at the temperatures indicated on the labels
For Use With (Application) RNA Binding Protein Immunoprecipitation (RIP), Protein Interaction Assays
Quantity 24 Assays
Stability Kit components are stable for 6 months from date of shipment when stored as directed
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.