Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ EZ-Magna Nuclear RIP™ (Native) Nuclear RNA-Binding Protein Immunoprecipitation Kit
Shop All MilliporeSigma Products
Click to view available options
Quantity:
24 Assays
Description
Includes
- Nuclei Isolation Buffer
- RIP Lysis Buffer
- Protein A/G Magnetic Beads
- RIP Lysis Buffer
- 0.5M EDTA
- DNase I (RNase Free) 2U/μL
- DNase I Reaction Buffer
- Protease Inhibitor Cocktail III, Animal Free
- RNAse Inhibitor
Requires
- Antibody of interest for RNA-binding protein immunoprecipitation (RIP)
- Negative Control Antibody
- Precipitation Carrier (e.g. Pellet Paint™ Co-Precipitant or linear Acrylamide (5mg/mL) if samples are intended for Next Gen Sequence library preparation
- Reagents for qRT-PCR Analysis and/or Reagents for RNA-Seq Library Construction (optional)
- Generates native chromatin to allow analysis of a variety of chromatin-associated RNAs
- Flexible, scalable input requirements: Recover RNA from millions of cells or as few as 100,000 cells (native)
- Magnetic protein A/G bead blend and optimized buffer system results in low backgrounds and high signal-to-noise ratios
- Suitable for analysis by RT-qPCR or RIP-seq
- Complete set of reagents and detailed protocol to enable first-time success
- Includes positive and negative control antibodies and qPCR primers

Specifications
Specifications
Content And Storage | Store components at the temperatures indicated on the labels |
For Use With (Application) | RNA Binding Protein Immunoprecipitation (RIP), Protein Interaction Assays |
Quantity | 24 Assays |
Stability | Kit components are stable for 6 months from date of shipment when stored as directed |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction