Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nucleocapsid Antibody (1051436), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB305297 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305297 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305297 Supplier Novus Biologicals Supplier No. FAB11289A
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nucleocapsid Monoclonal antibody specifically detects Nucleocapsid in HCoV-OC43 samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry

Specifications

Antigen Nucleocapsid
Applications Western Blot, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 1051436
Conjugate APC
Dilution Western Blot, Flow Cytometry, Immunocytochemistry
Formulation PBS
Host Species Mouse
Immunogen Sf-21 (baculovirus)-derived hcov-oc43 Nucleocapsid, Met1-Ile448, Accession # YP_009555245.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species HCoV-OC43
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.