Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nucleolin Antibody (NCL/7014R), PerCP, Novus Biologicals™
SDP

Catalog No. NB121348 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB121348 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB121348 Supplier Novus Biologicals Supplier No. NBP311038PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Nucleolin Monoclonal antibody specifically detects Nucleolin in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Nucleolin
Applications Flow Cytometry
Classification Monoclonal
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS
Gene Alias C23, FLJ45706, FLJ59041, nucleolin, Protein C23
Host Species Rabbit
Immunogen Recombinant full-length human Nucleolin protein
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4691
Target Species Human, Bovine (Negative), Mouse (Negative), Rat (Negative)
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.