Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nucleolin Antibody (NCL/7338), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB186941 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB186941 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB186941 Supplier Novus Biologicals Supplier No. NBP314014G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Nucleolin Monoclonal antibody specifically detects Nucleolin in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Nucleolin
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone NCL/7338
Conjugate DyLight 488
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C23, FLJ45706, FLJ59041, nucleolin, Protein C23
Host Species Mouse
Immunogen Recombinant full-length human Nucleolin protein (Uniprot: P19338)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4691
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.