Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nucleolin/NCL Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501558271
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-8271 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-8271 Supplier Bethyl Laboratories, Inc Supplier No. A300709AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict

Nucleolin, which is identical to human DNA helicase IV, is a major nucleolar phosphoprotein which is associated with preribosomal RNA and is implicated in the early stage of preribosomal RNP assembly and processing. This 100 kDa protein has three major domains: a N-terminal domain comprised of long acidic stretches interspersed with basic repeats, similar to the structure of a high mobility group-type protein, a central domain that contains four RNA binding elements, a C-terminal domain approximately 85 amino acids long that is rich in glycine, arginine, and phenylalanine residues. Nucleolin fluctuates in parallel to DNA synthesis; intact 100 kDa protein is the major species in actively dividing cells, whereas the degraded forms are relativley abundant in nondividing cells. Nucleolin can unwind RNA-RNA duplexes, as well as DNA-DNA and DNA-RNA duplexes. Nucleolin also interacts directly with DNA topoisomerase I. It is located mainly in dense fibrillar regions of the nucleolus. Nucleolin is the major nucleolar protein of growing eukaryotic cells. It is found associated with intranucleolar chromatin and preribosomal particles. It induces chromatin decondensation by binding to histone H1. It is thought to play a role in pre-rRNA transcription and ribosome assembly. It interacts with APTX and contains 4 RNA recognition motif (RRM) domains.

Specifications

Antigen Nucleolin/NCL
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene NCL
Gene Accession No. P19338
Gene Alias C23, FLJ45706, FLJ59041
Gene Symbols NCL
Host Species Rabbit
Immunogen Between 250 and 300
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4691
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.