Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nucleoporin NUP85 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001355 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001355 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001355 Supplier Novus Biologicals Supplier No. NB1001355
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Nucleoporin NUP85 Polyclonal specifically detects Nucleoporin NUP85 in Human samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen Nucleoporin NUP85
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_079120.
Gene Alias 85 kDa nucleoporin, FLJ12549, frount, nuclear pore complex protein Nup85, nucleoporin 85kDa, Nucleoporin Nup75, Nucleoporin Nup85, Nup75, PCNT, PCNT1, Pericentrin-1
Gene Symbols NUP85
Host Species Goat
Immunogen Peptide with sequence C-ARAIIREGSLEGS corresponding to C-Terminus according to NP_079120.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79902
Test Specificity Please note that, in contrast to other anti-Pericentrin antibodies available, this ab is NOT expected to recognize Kendrin (also known as Pericentrin 2). This may explain why it recognizes a 75KDa band (the expected size according to NP_079120) and not a 220kDa band (as reported for other anti-pericentrin antibodies).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.