Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nucleostemin Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB105447 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB105447 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB105447 Supplier Novus Biologicals Supplier No. NBP298743V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Nucleostemin Polyclonal specifically detects Nucleostemin in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Nucleostemin
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias C77032, E2IG3NNP47, estradiol-induced nucleotide binding protein, guanine nucleotide binding protein-like 3 (nucleolar), guanine nucleotide-binding protein-like 3, MGC800, Novel nucleolar protein 47, NSE2-induced gene 3 protein, Nucleolar GTP-binding protein 3, nucleostemin
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Nucleostemin.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cellular Markers, Core ESC Like Genes, Epigenetics, mTOR Pathway, Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 26354
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.