Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nucleostemin Antibody, Novus Biologicals™
SDP

Catalog No. NBP238326 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP238326 0.1 mL
NB436241 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP238326 Supplier Novus Biologicals Supplier No. NBP238326
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Nucleostemin Polyclonal specifically detects Nucleostemin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Nucleostemin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q9BVP2
Gene Alias C77032, E2IG3NNP47, estradiol-induced nucleotide binding protein, guanine nucleotide binding protein-like 3 (nucleolar), guanine nucleotide-binding protein-like 3, MGC800, Novel nucleolar protein 47, NSE2-induced gene 3 protein, Nucleolar GTP-binding protein 3, nucleostemin
Gene Symbols GNL3
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: SILFQSSGLTNGIIEEKDIHEELPKRKERKQEEREDDKDSDQETVDEEVDENSSGMFAAEETGEALSEETTAGEQSTRSFI
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cellular Markers, Chromatin Modifiers, Core ESC Like Genes, Embryonic Stem Cell Markers, Epigenetics, Histones and Modified Histones, Metabotropic Glutamate Receptors, microRNA, mTOR Pathway, Neurofilaments, Stem Cell Markers, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 26354
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.