Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nucleotide binding protein like Antibody (OTI5D5), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP273094A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP273094A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP273094A32 Supplier Novus Biologicals Supplier No. NBP273094AF532

Mouse Monoclonal Antibody

Nucleotide binding protein like Monoclonal specifically detects Nucleotide binding protein like in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Nucleotide binding protein like
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI5D5
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias C14orf127, FLJ12660, huInd1IND1 homolog, IND1chromosome 14 open reading frame 127, iron-sulfur protein NUBPL, iron-sulfur protein required for NADH dehydrogenase, nucleotide binding protein-like, Nucleotide-binding protein-like
Gene Symbols NUBPL
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-250 of human Nucleotide binding protein like (NP_079428) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 80224
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.