Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nudel Antibody (OTI5E11), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N273104APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273104APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273104APC7 Supplier Novus Biologicals Supplier No. NBP273104APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nudel Monoclonal specifically detects Nudel in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Nudel
Applications Flow Cytometry
Classification Monoclonal
Clone OTI5E11
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias DKFZp451M0318, EOPA, MITAP1mitosin-associated protein MITAP1, Mitosin-associated protein 1, nuclear distribution protein nudE-like 1, nudE nuclear distribution gene E homolog (A. nidulans)-like 1, nudE nuclear distribution gene E homolog like 1, nudE nuclear distribution gene E homolog like 1 (A. nidulans), NUDELendooligopeptidase A, NUDE-like protein, Protein Nudel
Gene Symbols NDEL1
Host Species Mouse
Immunogen Full length recombinant protein of human Nudel (NP_001020750) produced in HEk293 cell.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 81565.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.