Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT10 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB102111 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB102111 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB102111 Supplier Novus Biologicals Supplier No. NBP297464C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NUDT10 Polyclonal specifically detects NUDT10 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen NUDT10
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Formulation 50 mM Sodium Borate
Gene Alias APS2DIPP3A, Diadenosine 5'-5'''-P1, diphosphoinositol polyphosphate phosphohydrolase 3-alpha, diphosphoinositol polyphosphate phosphohydrolase type 3 alpha, DIPP3a, DIPP-3-alpha, DIPP3-alpha, EC 3.6.1.-, EC 3.6.1.52, hDIPP3alphahAps2, Nucleoside diphosphate-linked moiety X motif 10, nudix (nucleoside diphosphate linked moiety X)-type motif 10, Nudix motif 10, P6-hexaphosphate hydrolase 3-alpha
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human NUDT10 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 170685
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.