Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT10 Antibody (OTI1A6), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB273105A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB273105A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB273105A48 Supplier Novus Biologicals Supplier No. NBP273105AF488

Mouse Monoclonal Antibody

NUDT10 Monoclonal specifically detects NUDT10 in Human samples. It is validated for Western Blot.

Specifications

Antigen NUDT10
Applications Western Blot
Classification Monoclonal
Clone OTI1A6
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias APS2DIPP3A, Diadenosine 5′-5′′′-P1, diphosphoinositol polyphosphate phosphohydrolase 3-alpha, diphosphoinositol polyphosphate phosphohydrolase type 3 alpha, DIPP3a, DIPP-3-alpha, DIPP3-alpha, EC 3.6.1.-, EC 3.6.1.52, hDIPP3alphahAps2, Nucleoside diphosphate-linked moiety X motif 10, nudix (nucleoside diphosphate linked moiety X)-type motif 10, Nudix motif 10, P6-hexaphosphate hydrolase 3-alpha
Gene Symbols NUDT10
Host Species Mouse
Immunogen Full length recombinant protein of human NUDT10 (NP_694853) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 170685.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.