Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT10 Antibody (OTI1A6), PerCP, Novus Biologicals™
SDP

Catalog No. NB302205 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302205 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302205 Supplier Novus Biologicals Supplier No. NBP273105PCP

Mouse Monoclonal Antibody

NUDT10 Monoclonal antibody specifically detects NUDT10 in Human samples. It is validated for Western Blot

Specifications

Antigen NUDT10
Applications Western Blot
Classification Monoclonal
Clone OTI1A6
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias APS2DIPP3A, Diadenosine 5'-5'''-P1, diphosphoinositol polyphosphate phosphohydrolase 3-alpha, diphosphoinositol polyphosphate phosphohydrolase type 3 alpha, DIPP3a, DIPP-3-alpha, DIPP3-alpha, EC 3.6.1.-, EC 3.6.1.52, hDIPP3alphahAps2, Nucleoside diphosphate-linked moiety X motif 10, nudix (nucleoside diphosphate linked moiety X)-type motif 10, Nudix motif 10, P6-hexaphosphate hydrolase 3-alpha
Host Species Mouse
Immunogen Full length human recombinant protein of human NUDT10 (NP_694853) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 170685
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.