Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUDT11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113172
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113172 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113172 Supplier Invitrogen™ Supplier No. PA5113172
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NUDT11 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen NUDT11
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene NUDT11
Gene Accession No. Q96G61
Gene Alias APS1; ASP1; Diadenosine 5',5'''-P1,P6-hexaphosphate hydrolase 3-alpha; Diadenosine 5',5'''-P1,P6-hexaphosphate hydrolase 3-beta; diadenosine hexaphosphate hydrolase (AMP-forming); diphosphoinositol polyphosphate phosphohydrolase 3, beta; Diphosphoinositol polyphosphate phosphohydrolase 3-alpha; Diphosphoinositol polyphosphate phosphohydrolase 3-beta; DIPP3; DIPP3 beta; Dipp3a; DIPP-3-alpha; Dipp3b; DIPP3beta; DIPP-3-beta; DIPP3-beta; hAps1; hDIPP3beta; MNCb-1696; Nucleoside diphosphate-linked moiety X motif 10; Nucleoside diphosphate-linked moiety X motif 11; nudix (nucleoside diphosphate linked moiety X)-type motif 11; nudix hydrolase 11; Nudix motif 10; Nudix motif 11; nudix-type motif 11; Nudt10; Nudt11
Gene Symbols NUDT11
Host Species Rabbit
Immunogen Recombinant Human Diphosphoinositol polyphosphate phosphohydrolase 3-beta protein (1-164AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55190
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.