Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT16 Antibody, Novus Biologicals™
SDP

Catalog No. NBP237992 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237992 0.1 mL
NB436346 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP237992 Supplier Novus Biologicals Supplier No. NBP237992
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NUDT16 Polyclonal specifically detects NUDT16 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen NUDT16
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q96DE0
Gene Alias EC 3.6.1, EC 3.6.1.-, FLJ31265, FLJ34034, FLJ36248, Nucleoside diphosphate-linked moiety X motif 16, nudix (nucleoside diphosphate linked moiety X)-type motif 16, Nudix motif 16, U8 snoRNA-binding protein H29K, U8 snoRNA-decapping enzyme
Gene Symbols NUDT16
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: SAREQLLEALQDLGLLQSGSISGLKIPAHH
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 131870
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.