Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT16L1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP155432 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP155432 100 μL
NBP15543220 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP155432 Supplier Novus Biologicals Supplier No. NBP155432
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NUDT16L1 Polyclonal specifically detects NUDT16L1 in Human samples. It is validated for Western Blot.

Specifications

Antigen NUDT16L1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9BRJ7
Gene Alias MGC11275, nudix (nucleoside diphosphate linked moiety X)-type motif 16-like 1, protein syndesmos, SDOSNUDT16-like protein 1
Gene Symbols NUDT16L1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to NUDT16L1(nudix (nucleoside diphosphate linked moiety X)-type motif 16-like 1) The peptide sequence was selected from the C terminal of NUDT16L1. Peptide sequence GLVRVPLYTQKDRVGGFPNFLSNAFVSTAKCQLLFALKVLNMMPEEKLVE The peptide sequence for this immunogen was taken from within the described region.
Molecular Weight of Antigen 23 kDa
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 84309
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.