Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT18 Antibody (OTI5D7), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB273110APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB273110APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB273110APC Supplier Novus Biologicals Supplier No. NBP273110APC

Mouse Monoclonal Antibody

NUDT18 Monoclonal specifically detects NUDT18 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen NUDT18
Applications Flow Cytometry
Classification Monoclonal
Clone OTI5D7
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 3.6.1, EC 3.6.1.-, FLJ22494, nucleoside diphosphate-linked moiety X motif 18, nudix (nucleoside diphosphate linked moiety X)-type motif 18, Nudix motif 18
Gene Symbols NUDT18
Host Species Mouse
Immunogen Full length recombinant protein of human NUDT18 (NP_079091) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 79873.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.