Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT18 Antibody (OTI5D7), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB281349 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB281349 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB281349 Supplier Novus Biologicals Supplier No. NBP273110MFV450

Mouse Monoclonal Antibody

NUDT18 Monoclonal antibody specifically detects NUDT18 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen NUDT18
Applications Flow Cytometry
Classification Monoclonal
Clone OTI5D7
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias EC 3.6.1, EC 3.6.1.-, FLJ22494, nucleoside diphosphate-linked moiety X motif 18, nudix (nucleoside diphosphate linked moiety X)-type motif 18, Nudix motif 18
Host Species Mouse
Immunogen Full length human recombinant protein of human NUDT18(NP_079091) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 79873
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.