Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT21 Antibody (OTI13H1), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP273113V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273113V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273113V Supplier Novus Biologicals Supplier No. NBP273113V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NUDT21 Monoclonal specifically detects NUDT21 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NUDT21
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI13H1
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CFIm25, CFIM25CPSF 25 kDa subunit, cleavage and polyadenylation specific factor 5, 25 kD subunit, cleavage and polyadenylation specific factor 5, 25 kDa, Cleavage and polyadenylation specificity factor 25 kDa subunit, cleavage and polyadenylation specificity factor subunit 5, CPSF25, CPSF5DKFZp686H1588, Nucleoside diphosphate-linked moiety X motif 21, nudix (nucleoside diphosphate linked moiety X)-type motif 21, Nudix motif 21, pre-mRNA cleavage factor Im (25kD), Pre-mRNA cleavage factor Im 25 kDa subunit, pre-mRNA cleavage factor Im, 25kD subunit
Gene Symbols NUDT21
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 2-227 of human NUDT21 (NP_008937) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 11051.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.