Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT6 Antibody (OTI9D12), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N273121PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273121PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273121PEC5 Supplier Novus Biologicals Supplier No. NBP273121PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NUDT6 Monoclonal specifically detects NUDT6 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen NUDT6
Applications Flow Cytometry
Classification Monoclonal
Clone OTI9D12
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Antisense basic fibroblast growth factor, ASFGF2, bFGF, FGF2ASFGF-2, FGF-AS, gfg, gfg-1, nucleoside diphosphate-linked moiety X motif 6, nudix (nucleoside diphosphate linked moiety X)-type motif 6, Nudix motif 6, nudix-type motif 6, Protein GFG
Gene Symbols NUDT6
Host Species Mouse
Immunogen Full length recombinant protein of human NUDT6 (NP_009014) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 11162.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.