Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT9 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB354332 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB354332 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB354332 Supplier Novus Biologicals Supplier No. NBP297230MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NUDT9 Polyclonal antibody specifically detects NUDT9 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen NUDT9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ADP-ribose diphosphatase, ADP-ribose phosphohydrolase, ADP-ribose pyrosphosphatase NUDT9, ADPR-PPase, EC 3.6.1.13, MGC3037, mitochondrial, nucleoside diphosphate linked moiety X-type motif 9, nudix (nucleoside diphosphate linked moiety X)-type motif 9, Nudix motif 9, NUDT10
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human NUDT9 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 53343
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.