Learn More
NUF2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304319AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Nuf2 (also known as CDCA1 or cell division cycle associated 1) is important for the formation of stable kinetochore-microtubule attachments, chromosome alignment in mitosis, and mitotic progression. Nuf2 is a spindle pole body-associated protein required for nuclear division, spindle pole body separation, and spindle elongation. Nuf2 is part of the Ndc80 complex, which is also comprised of Ndc80 (Hec1), Spc24, and Spc25. The Ndc80 complex is essential for metaphase chromosome alignment and anaphase chromosome segregation.Specifications
NUF2 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q99P69, Q9BZD4 | |
Nuf2 | |
Between 414 and 464 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
NUF2 | |
CDCA1, CT106, NUF2R, cancer/testis antigen 106, cell division cycle associated 1, cell division cycle-associated protein 1, hNuf2, hNuf2R, hsNuf2, kinetochore protein Nuf2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
66977, 83540 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.