Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUF2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | NUF2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-7421
![]() |
Bethyl Laboratories, Inc
A304319AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
Nuf2 (also known as CDCA1 or cell division cycle associated 1) is important for the formation of stable kinetochore-microtubule attachments, chromosome alignment in mitosis, and mitotic progression. Nuf2 is a spindle pole body-associated protein required for nuclear division, spindle pole body separation, and spindle elongation. Nuf2 is part of the Ndc80 complex, which is also comprised of Ndc80 (Hec1), Spc24, and Spc25. The Ndc80 complex is essential for metaphase chromosome alignment and anaphase chromosome segregation.Specifications
NUF2 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
CDCA1, CT106, NUF2R, cancer/testis antigen 106, cell division cycle associated 1, cell division cycle-associated protein 1, hNuf2, hNuf2R, hsNuf2, kinetochore protein Nuf2 | |
Between 414 and 464 | |
Primary | |
4° C | |
NUF2 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q99P69, Q9BZD4 | |
66977, 83540 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title