Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUFIP2 Antibody, Novus Biologicals™
SDP

Catalog No. NB10079842 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10079842 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10079842 Supplier Novus Biologicals Supplier No. NB10079842
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NUFIP2 Polyclonal specifically detects NUFIP2 in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen NUFIP2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 mcg/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q7Z417
Gene Alias 182-FIP, Cell proliferation-inducing gene 1 protein, FIP-82, FLJ10976, FMRP-interacting protein 2, Fragile X Mental Retardation Protein, KIAA1321PIG1,82-FIP82-kD FMRP Interacting Protein, MGC117262, nuclear fragile X mental retardation protein interacting protein 2, nuclear fragile X mental retardation-interacting protein 2, proliferation-inducing gene 1,82 kDa FMRP-interacting protein
Gene Symbols NUFIP2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 645 and 695 of human nuclear fragile X mental retardation protein interacting protein 2 (82 kDa FMRP-interacting protein) using the numbering given in entry NP_065823.1 (GeneID 575
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 57532
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, African elephant, Naked mole rat, Small-eared galago and Northern white-cheeked gibbon.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.