Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUFIP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094585 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094585 20 μL
NB094584 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094585 Supplier Novus Biologicals Supplier No. NBP30505120UL

Rabbit Polyclonal Antibody

NUFIP2 Polyclonal specifically detects NUFIP2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen NUFIP2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias 182-FIP, Cell proliferation-inducing gene 1 protein, FIP-82, FLJ10976, FMRP-interacting protein 2, Fragile X Mental Retardation Protein, KIAA1321PIG1,82-FIP82-kD FMRP Interacting Protein, MGC117262, nuclear fragile X mental retardation protein interacting protein 2, nuclear fragile X mental retardation-interacting protein 2, proliferation-inducing gene 1,82 kDa FMRP-interacting protein
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 456-695 of human NUFIP2 (NP_065823.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 57532
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.