Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NULP1 Antibody (PCRP-TCF25-1F12), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB381226 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB381226 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB381226 Supplier Novus Biologicals Supplier No. NBP308354DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NULP1 Monoclonal antibody specifically detects NULP1 in Human samples. It is validated for Western Blot, Protein Array

Specifications

Antigen NULP1
Applications Western Blot, Protein Array
Classification Monoclonal
Clone PCRP-TCF25-1F12
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias hKIAA1049, KIAA1049Hulp1, Nuclear localized protein 1, Nulp1, NULP1PRO2620, TCF-25, transcription factor 25, transcription factor 25 (basic helix-loop-helix)
Host Species Mouse
Immunogen Recombinant full-length human NULP1 protein (Uniprot: Q9BQ70)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22980
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.