Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NuMA Antibody, Novus Biologicals™
SDP

Catalog No. NB500174 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500174 0.1 mL
NB500174SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB500174 Supplier Novus Biologicals Supplier No. NB500174
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 18 publications

NuMA Polyclonal specifically detects NuMA in Human, Mouse, Rat, Mammal, Marsupial, Primate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen NuMA
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Simple Western 1:500, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml, Immunohistochemistry-Frozen reported in scientific literature
Formulation Whole antisera with 0.02% Sodium Azide
Gene Accession No. Q14980
Gene Alias centrophilin stabilizes mitotic spindle in mitotic cells, nuclear mitotic apparatus protein 1, NUMA, NuMA protein, SP-H antigen, structural nuclear protein
Gene Symbols NUMA1
Host Species Rabbit
Immunogen Full-length recombinant human NuMA expressed in insect Sf9 cells using baculovirus expression system, then purified to near homogeneity prior to immunization.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Mitotic Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4926
Test Specificity Reactive to most mammalian species tested.
Target Species Human, Mouse, Rat, Mammalia, Marsupial, Primate
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.