Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NuMA Antibody (12A11), Novus Biologicals™
SDP

Catalog No. NB298074 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298074 50 μL
NB298075 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298074 Supplier Novus Biologicals Supplier No. NBP32645150UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

NuMA Monoclonal antibody specifically detects NuMA in Human samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen NuMA
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 12A11
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:200, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias centrophilin stabilizes mitotic spindle in mitotic cells, nuclear mitotic apparatus protein 1, NUMA, NuMA protein, SP-H antigen, structural nuclear protein
Host Species Rabbit
Immunogen A synthesized peptide derived from Human NuMA [UniProt Q14980]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Breast Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Mitotic Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4926
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.