Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NuMA Antibody (12A11), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | NuMA |
---|---|
Clone | 12A11 |
Dilution | Flow Cytometry 1:50-1:200, ELISA |
Applications | Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
NuMA Monoclonal antibody specifically detects NuMA in Human samples. It is validated for Flow Cytometry, ELISASpecifications
NuMA | |
Flow Cytometry 1:50-1:200, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
centrophilin stabilizes mitotic spindle in mitotic cells, nuclear mitotic apparatus protein 1, NUMA, NuMA protein, SP-H antigen, structural nuclear protein | |
A synthesized peptide derived from Human NuMA [UniProt Q14980] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
12A11 | |
Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Mitotic Regulators | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
4926 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title