Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Numb Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB500178Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500178Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500178Y Supplier Novus Biologicals Supplier No. NB500178AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Numb Polyclonal specifically detects Numb in Human, Mouse, Chicken samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Numb
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias h-Numb, numb (Drosophila) homolog, numb homolog (Drosophila), protein numb homolog, Protein S171, S171
Gene Symbols NUMB
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of mouse NUMB (between residues 600-653). [UniProt# Q9QZS3]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8650
Test Specificity This is specific for all four isoforms of the NUMB protein.
Target Species Human, Mouse, Chicken
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.