Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Numb Antibody (OTI1B5), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP273130C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273130C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273130C Supplier Novus Biologicals Supplier No. NBP273130C

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Numb Monoclonal specifically detects Numb in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Numb
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1B5
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias h-Numb, numb (Drosophila) homolog, numb homolog (Drosophila), protein numb homolog, Protein S171, S171
Gene Symbols NUMB
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-365 of human Numb (NP_001005744) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8650.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.