Learn More
Thermo Scientific™ Nunc™ 384-Shallow Well Standard Height Polypropylene Sample Processing & Storage Microplates
GREENER_CHOICE
Catalog No. 12566213
Click to view available options
Color:
Black
Natural
White
Sterility:
Non-sterile
Sterile
Fit for Purpose:
Certified Quality
Lower spending when storing compounds for small volume applications by using these 384-shallow well standard height polypropylene microplates.
Lower spending when storing compounds for small volume applications by using Thermo Scientific™ Nunc™ 384-Shallow Well Standard Height Polypropylene Microplates. Polypropylene is ideal for storage plates because it has a lower binding capacity to prevent proteins or DNA binding, and allows for complete sample recovery. Withstands temperatures from -80° to +121°C. Offered in a variety of colors to allow for quick identification of storage plates.
Features:
  • Rounded wells are optimized for mixing and sample recovery
  • Square rounded wells minimize wicking (capillary action)
  • Natural plates for storage identification or storing colorimetric assays
  • White plates for luminescent assays
  • Black plates for fluorescent assays
  • Customized barcoding is available
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Certifications/Compliance Certified RNase and DNase Free. ANSI Compliant.
Format Standard
Lid Without Lid
Description 384-well Polypropylene Sample Processing & Storage Plates
Array 16 x 24
Autoclavable Autoclavable
Packaging 100 Cases
Binding Type Low binding PP
Product Line Nunc
Color White
Product Type 384-well plate
For Use With (Equipment) ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
Material Polypropylene
Volume (Metric) Well 55 μL
No. of Wells 384
Volume (Metric) Working 2 to 35 μL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Square, Round Bottom (U-well)
Dnase Free Yes
For Use With (Application) Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
No. per Case 100
No. per Pack 25
Regulatory Status For Research Use Only. Not for use in diagnostic procedures.
RNase Free Yes
Fit for Purpose Certified Quality
Plate Size ANSI/ SLAS Standard
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.