Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ Seals, Tapes, and Foils
GSA_VAGREENER_CHOICE

Catalog No. 12566914
Click to view available options
Material:
Aluminum
Non-woven Rayon
PVC-coated Polypropylene
Plastic
Polyester
Polyethylene
Polyolefin
Polypropylene
Vinyl
For Use With (Application):
Breathable, Cell culture
Cell culture, sterile applications
Cell growth, Bacterial growth
Consistent seal
Crystallography and genotyping
General purpose, ELISA
General purpose, sample detection
Low autofluorescence, ELISA
Luminescence, photosensitive samples
PCR
PCR, fluorescence assays, microscopy
PCR, long-term storage
PCR, qPCR, ELISA, Protein crystallography
PCR, storage in 384 well plates
PCR, storage in 96-well plates
Short-term storage, ELISA
Short-term storage, ELISA, minimizes evaporation and cross contamination
Ventilating and storing cell cultures
Certified RNase/DNase-free:
No
Yes

Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.

Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.

  • Adhesive seals effectively seal all microplate formats
  • Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
  • Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Solvent Or Alcohol Resistant Life Science Reagent Resistant
Description Sealing Tape, Polyester, Clear, Roll
Material Polyester
Sterility Non-sterile
Autoclavable Not Autoclavable
No. of Wells 96
No. per Case 1000
No. per Pack 1000
Evaporation Barrier Minimizes evaporation and cross contamination
For Use With (Application) General purpose, ELISA
Content And Storage General
Product Type Adhesive Seal
Product Line Nunc
Adhesive Acrylic
Certified RNase/DNase-free Yes
Pierceable No
Temperature Range (Metric) -40°C to +120°C
Well Shape All well shapes
Plate Compatibility All Microplates
Incubation Conditions General
Peelable Yes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.