Learn More
Description
Specifications
Specifications
| Antigen | NUP133 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | FLJ10814, hNUP133,133 kDa nucleoporin, MGC21133, nuclear pore complex protein Nup133, nucleoporin 133kD, nucleoporin 133kDa, Nucleoporin Nup133 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human NUP133 (NP_060700.2). Peptide sequence QVLRDAPMDSIEWAEVVINVNNILKDMLQAASHYRQNRNSLYRREESLEK |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
