Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUP160 Antibody, Novus Biologicals™
SDP

Catalog No. NBP223718 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223718 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223718 Supplier Novus Biologicals Supplier No. NBP223718
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NUP160 Polyclonal specifically detects NUP160 in Human, Bovine, Equine, Primate samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen NUP160
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1:100
Formulation 0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide
Gene Accession No. Q12769
Gene Alias DKFZp686M14102, FLJ22583, KIAA0197MGC150678, MGC150679,160 kDa nucleoporin, nuclear pore complex protein Nup160, nucleoporin 160kD, nucleoporin 160kDa, Nucleoporin Nup160, NUP120
Gene Symbols NUP160
Host Species Rabbit
Immunogen Within the range of amino acids 668-699 of human NUP160 protein were used as the immunogen.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23279
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_c) and chimpanzee, 94% homologous in rhesus monkey, 88% homologous in horse and 83% homologous in cow.
Target Species Human, Bovine, Equine, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.