Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUP205 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB290305 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290305 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290305 Supplier Novus Biologicals Supplier No. NBP222287PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NUP205 Polyclonal antibody specifically detects NUP205 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen NUP205
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias nucleoporin 205kDa
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1750 and 1800 of human Nucleoporin 205kDa using the numbering given in entry NP_055950.1 (GeneID 23165).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23165
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.