Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUP37 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP174146
Description
NUP37 Polyclonal specifically detects NUP37 in Mouse samples. It is validated for Western Blot.Specifications
| NUP37 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| MGC5585, nucleoporin 37kDa, nucleoporin Nup37, Nup107-160 subcomplex subunit Nup37, p37FLJ22618 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79023 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9CWU9 | |
| NUP37 | |
| Synthetic peptides corresponding to the N terminal of Nup37. Immunizing peptide sequence EEETDIEGIQYKTLRTFHHGVRVDGIAWSPETKLDSLPPVIKFCTSAADL. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Chicken: 92%; Canine: 92%; Pig: 85%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction