Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUP37 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | NUP37 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
NUP37 Polyclonal specifically detects NUP37 in Mouse samples. It is validated for Western Blot.Specifications
| NUP37 | |
| Polyclonal | |
| Rabbit | |
| Q9CWU9 | |
| 79023 | |
| Synthetic peptides corresponding to the N terminal of Nup37. Immunizing peptide sequence EEETDIEGIQYKTLRTFHHGVRVDGIAWSPETKLDSLPPVIKFCTSAADL. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| MGC5585, nucleoporin 37kDa, nucleoporin Nup37, Nup107-160 subcomplex subunit Nup37, p37FLJ22618 | |
| NUP37 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title