Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUP43 Antibody (OTI6F8), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP273131G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273131G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273131G Supplier Novus Biologicals Supplier No. NBP273131G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NUP43 Monoclonal specifically detects NUP43 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen NUP43
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI6F8
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias bA350J20.1, FLJ13287, nucleoporin 43kDa, nucleoporin Nup43, Nup107-160 subcomplex subunit Nup43, p42
Gene Symbols NUP43
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-253 of human NUP43 (NP_942590) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 348995.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.