Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUP50 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB102033 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB102033 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB102033 Supplier Novus Biologicals Supplier No. NBP297442G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NUP50 Polyclonal specifically detects NUP50 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen NUP50
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Formulation 50 mM Sodium Borate
Gene Alias 50 kDa nucleoporin, MGC39961, NPAP60, NPAP60LNucleoporin Nup50, nuclear pore complex protein Nup50, Nuclear pore-associated protein 60 kDa-like, nuclear pore-associated protein 60L, nucleoporin 50kD, nucleoporin 50kDa
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human NUP50 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10762
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.