Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nup53 Antibody, Novus Biologicals™
SDP

Catalog No. NB10093322 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093322 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10093322 Supplier Novus Biologicals Supplier No. NB10093322
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Nup53 Polyclonal specifically detects Nup53 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Nup53
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:250- 1:1000, Immunoprecipitation 2 - 10 ug/mg lysate, Immunohistochemistry-Paraffin 1:500 - 1:2000
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_612142.2
Gene Alias 35 kDa nucleoporin, Mitotic phosphoprotein 44, MP-44, MP44NUP53, NP44, Nuclear pore complex protein Nup53, nucleoporin 35kDa, Nucleoporin Nup35, nucleoporin NUP53
Gene Symbols NUP35
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 276 and 376 of human nucleoporin 35kDa using the numbering given in entry NP_612142.2 (GeneID 129401).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129401
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Whitetufted-earmarmoset, Crab-eating macaque, European domestic ferret and Northern white-cheekedgibbon.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.