Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUP62 Antibody (HL1226) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB295036 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295036 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295036 Supplier Novus Biologicals Supplier No. NBP325581
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

NUP62 Monoclonal antibody specifically detects NUP62 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen NUP62
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HL1226
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias DKFZp547L134,62 kDa nucleoporin, FLJ20822, FLJ43869, IBSNp62, MGC841, nuclear pore glycoprotein p62, nucleoporin 62kD, nucleoporin 62kDa, SNDINucleoporin Nup62
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NUP62. The exact sequence is proprietary.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23636
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.