Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUP62 Antibody (RL31), HRP, Novus Biologicals™
SDP

Catalog No. NB359552 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359552 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359552 Supplier Novus Biologicals Supplier No. NB1202736H
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

NUP62 Monoclonal antibody specifically detects NUP62 in Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen NUP62
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone RL31
Conjugate HRP
Dilution Western Blot, Immunofluorescence
Formulation PBS
Gene Alias DKFZp547L134,62 kDa nucleoporin, FLJ20822, FLJ43869, IBSNp62, MGC841, nuclear pore glycoprotein p62, nucleoporin 62kD, nucleoporin 62kDa, SNDINucleoporin Nup62
Host Species Armenian Hamster
Immunogen O-linked glycoprotein-enriched fraction from rat liver nuclear envelopes raised in Armenian hamsters.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23636
Target Species Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.