Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUP88 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. p-7212218 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6173 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6173 Supplier Proteintech Group Inc Supplier No. 554651AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is specific to NUP88.

The nuclear pore complex (NPC) mediates bidirectional macromolecular traffic between the nucleus and cytoplasm in eukaryotic cells and is comprised of more than 100 different subunits. Many of the subunits belong to a family called nucleoporins (Nups), which are characterized by the presence of O-linked-N-acetylglucosamine moieties and a distinctive pentapeptide repeat (XFXFG). Nup88 (nucleoporin 88 kDa) is a 741 amino acid protein that localizes to the nucleus and functions as an essential component of the nuclear pore complex. Expressed ubiquitously, Nup88 is subject to phosphorylation by ATM or ATR and is upregulated in maligt neoplasms and precancerous dysplasias, suggesting a role in tumorigenesis. The gene encoding Nup88 maps to human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1, 200 genes.

Specifications

Antigen NUP88
Applications Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NUP88
Gene Accession No. Q99567
Gene Alias 88 kDa nucleoporin, nucleoporin 88kDa, Nucleoporin Nup88, NUP88
Gene Symbols NUP88
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4927
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.