Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUPL1 Antibody, Novus Biologicals™
SDP

Catalog No. NB438355 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB438355 25ul
NBP213684 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB438355 Supplier Novus Biologicals Supplier No. NBP21368425UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NUPL1 Polyclonal specifically detects NUPL1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NUPL1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25-2 ug/mL
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias KIAA0410PRO2463, nucleoporin like 1, nucleoporin p58/p45, Nucleoporin-like protein 1
Gene Symbols NUP58
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the amino acids: RNTLNIDKLKIETALELKNAEIALRTQKTPPGLQHEYAAPADYFRILVQQFEVQLQQYRQQIEELENHLATQANNSHITPQDLSMAMQKI
Purification Method Affinity Purified
Quantity 25ul
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9818
Test Specificity Specificity of human NUPL1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.