Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nurr1/NGFI-B beta/NR4A2 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB210652 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB210652 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB210652 Supplier Novus Biologicals Supplier No. NB11040415AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Nurr1/NGFI-B beta/NR4A2 Polyclonal antibody specifically detects Nurr1/NGFI-B beta/NR4A2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen Nurr1/NGFI-B beta/NR4A2
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias human homolog of, nuclear receptor subfamily 4, group A, member 2, orphan nuclear receptor NR4A2, Orphan nuclear receptor NURR1, T-cell nuclear receptor NOT, transcriptionally inducible nuclear receptor related 1, Transcriptionally-inducible nuclear receptor
Host Species Rabbit
Immunogen Synthetic peptide comprising residues 513-526 [TERHGLKEPKRVEE] of the human Nurr1 protein. Based on 100% sequence homology, this antibody is also expected to cross react with rat and mouse Nurr1.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immune System Diseases, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4929
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.